5-Bromo-2-chloro-3,4-pyridinediamine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloropyridine-3,4-diamine

A255069-10G A255069-1G A255069-250MG A255069-25G A255069-5G
76976-768EA 245.07 USD
76976-768 76976-770 76976-772 76976-774 76976-776
5-Bromo-2-chloro-3,4-pyridinediamine 97%
5-Bromo-2-chloro-3,4-pyridinediamine

Precaución: for Research Use Only

Formula: C₅H₅BrClN₃
MW: 222.48 g/mol
MDL Number: MFCD02266941
CAS Number: 163452-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR