3-Bromo-4-methoxy-5-nitropyridine 97%

Supplier: Ambeed
Warning

A367994-100MG A367994-1G A367994-250MG A367994-5G
76978-654EA 9 USD
76978-654 76978-656 76978-658 76978-660
3-Bromo-4-methoxy-5-nitropyridine 97%
3-Bromo-4-methoxy-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₃
MDL Number: MFCD16619117
CAS Number: 31872-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR