Guanosine-5'-diphosphate disodium salt 90% (by HPLC)
Supplier: Ambeed
|
Warning
|
Synonyms:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate, disodium salt, Guanosine 5'-diphosphate disodium salt
A119884-1G
A119884-250MG
A119884-25G
A119884-10G
A119884-5G
76971-432EA
68.96
USD
76971-432
76971-434
77611-014
77670-696
77670-698
Guanosine-5'-diphosphate disodium salt 90% (by HPLC)
Guanosine 5'-diphosphate disodium salt
Precaución: for Research Use Only
Formula:
C₁₀H₁₃N₅Na₂O₁₁P₂ MW: 487.17 g/mol |
MDL Number:
MFCD00070121 CAS Number: 7415-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...