2'-Fluoro-2'-deoxyguanosine 95%

Supplier: Ambeed
Warning

Synonyms: 2'-Deoxy-2'-fluoroguanosine, 2′-Deoxy-2′-fluoroguanosine, 2-Amino-9-((2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one

A194288-100MG A194288-1G A194288-250MG
76974-992EA 50.93 USD
76974-992 76974-994 76974-996
2'-Fluoro-2'-deoxyguanosine 95%
2'-Fluoro-2'-deoxyguanosine

Precaución: for Research Use Only

Formula: C₁₀H₁₂FN₅O₄
MW: 285.235 g/mol
MDL Number: MFCD00923832
CAS Number: 78842-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR