2'-Fluoro-2'-deoxyadenosine 98%

Supplier: Ambeed
Warning

Synonyms: 2′-Deoxy-2′-fluoroadenosine, (2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, 2'-Deoxy-2'-fluoroadenosine

A849783-100MG A849783-1G A849783-5G A849783-250MG
76981-792EA 27.59 USD
76981-792 76981-794 76981-796 77674-254
2'-Fluoro-2'-deoxyadenosine 98%
2'-Fluoro-2'-deoxyadenosine

Precaución: for Research Use Only

Formula: C₁₀H₁₂FN₅O₃
MW: 269.236 g/mol
MDL Number: MFCD09750859
CAS Number: 64183-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR