Cetrimonium bromide 98%

Supplier: Ambeed
Danger

A119034-25G A119034-500G A119034-100G
76971-392EA 14 USD
76971-392 77571-124 77610-990
Cetrimonium bromide 98%
Cetrimonium bromide

Precaución: for Research Use Only

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number: MFCD00011772
CAS Number: 57-09-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR