(2R,3R,4R,5R)-2-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2'-OMe-G(ibu) Phosphoramidite
A963776-1G
A963776-250MG
A963776-100MG
A963776-5G
76982-024EA
88
USD
76982-024
76982-026
77674-418
77882-850
(2R,3R,4R,5R)-2-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 95%
(2R,3R,4R,5R)-2-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
Precaución: for Research Use Only
Formula:
C₄₅H₅₆N₇O₉P MW: 869.94 g/mol |
MDL Number:
MFCD00792625 CAS Number: 150780-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...