5'-O-(4,4'-Dimethoxytrityl)-N-isobutyryl-2'-deoxyguanosine-3'-(2-cyanoethyl-N,N-diisopropyl)phosphoramidite 95%

Supplier: Ambeed
Warning

Synonyms: DMT-dG(ib) Phosphoramidite

A850305-10G A850305-1G A850305-5G
76981-798EA 119 USD
76981-798 76981-800 77882-772
5'-O-(4,4'-Dimethoxytrityl)-N-isobutyryl-2'-deoxyguanosine-3'-(2-cyanoethyl-N,N-diisopropyl)phosphoramidite 95%
5'-O-(4,4'-Dimethoxytrityl)-N-isobutyryl-2'-deoxyguanosine-3'-(2-cyanoethyl-N,N-diisopropyl)phosphoramidite

Precaución: for Research Use Only

Formula: C₄₄H₅₄N₇O₈P
MW: 839.91 g/mol
MDL Number: MFCD00055147
CAS Number: 93183-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR