Tacrolimus monohydrate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
FK 506, (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-Hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-8-(2-propen-1-yl)-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone monohydrate
A377737-10MG
A377737-1G
A377737-1MG
A377737-50MG
A377737-5MG
A377737-100MG
A377737-250MG
76978-808EA
17
USD
76978-808
76978-810
76978-812
76978-814
77613-136
77672-872
77672-874
Tacrolimus monohydrate 98%
Tacrolimus monohydrate
Precaución: for Research Use Only
Formula:
C₄₄H₇₁NO₁₃ MW: 822.03 g/mol |
MDL Number:
MFCD11045918 CAS Number: 109581-93-3 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...