Methyl-6-bromo-5-fluoronicotinate 98%

Supplier: Ambeed
Warning

A267342-5G A267342-1G A267342-250MG
76977-136EA 89 USD
76977-136 77612-568 77881-732
Methyl-6-bromo-5-fluoronicotinate 98%
Methyl-6-bromo-5-fluoronicotinate

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD14698110
CAS Number: 1214336-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR