Benzyltrimethylammonium chloride 97%

Supplier: Ambeed
Danger

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride

A846508-100G A846508-25G A846508-500G
76981-782EA 11 USD
76981-782 76981-784 76981-786
Benzyltrimethylammonium chloride 97%
Benzyltrimethylammonium chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆ClN
MDL Number: MFCD00011782
CAS Number: 56-93-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR