4-Bromo-2,6-difluorobenzotrifluoride 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1,3-difluoro-2-(trifluoromethyl)benzene

A224310-10G A224310-1G A224310-250MG A224310-25G A224310-5G
76975-922EA 114.58 USD
76975-922 76975-924 76975-926 76975-928 76975-930
4-Bromo-2,6-difluorobenzotrifluoride 97%
4-Bromo-2,6-difluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂BrF₅
MW: 260.99 g/mol
MDL Number: MFCD01631616
CAS Number: 156243-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR