5-Bromo-1-chloro-3-fluoro-2-methylbenzene 97%

Supplier: Ambeed
Warning

A345723-100MG A345723-1G A345723-250MG
76978-458EA 9 USD
76978-458 76978-460 76978-462
5-Bromo-1-chloro-3-fluoro-2-methylbenzene 97%
5-Bromo-1-chloro-3-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
MDL Number: MFCD21603712
CAS Number: 1806058-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR