2-Chloro-1,4-difluoro-3-methylbenzene 97%
Supplier: Ambeed
|
Warning
|
A259270-1G
A259270-250MG
76976-886EA
103
USD
76976-886
76976-888
2-Chloro-1,4-difluoro-3-methylbenzene 97%
2-Chloro-1,4-difluoro-3-methylbenzene
Precaución: for Research Use Only
Formula:
C₇H₅ClF₂ MW: 162.56 g/mol |
MDL Number:
MFCD12922611 CAS Number: 90292-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...