1-Bromo-2,5-difluoro-3-methylbenzene 98%

Supplier: Ambeed
Warning

A838320-1G A838320-250MG A838320-5G
76981-772EA 30 USD
76981-772 76981-774 77882-762
1-Bromo-2,5-difluoro-3-methylbenzene 98%
1-Bromo-2,5-difluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MDL Number: MFCD21603709
CAS Number: 1416354-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR