L(-)-Cystine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
A160767-1000G
A160767-100G
A160767-25G
A160767-500G
76973-586EA
93
USD
76973-586
76973-588
76973-590
76973-592
L(-)-Cystine 98%
L(-)-Cystine
Precaución: for Research Use Only
Formula:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH MW: 240.3 g/mol Boiling Pt: 428 |
MDL Number:
MFCD00064228 CAS Number: 56-89-3 Merck Index: 13,02811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...