3,5-Dibromo-4-methoxypyridine 98%

Supplier: Ambeed
Warning

A216027-1G A216027-250MG A216027-25G A216027-5G
76975-692EA 40.32 USD
76975-692 76975-694 76975-696 76975-698
3,5-Dibromo-4-methoxypyridine 98%
3,5-Dibromo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C6H5Br2NO
MW: 266.920 g/mol
MDL Number: MFCD13185524
CAS Number: 25813-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR