3-Cyano-2-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A217502-250MG A217502-1G A217502-5G
76975-740EA 27 USD
76975-740 77568-106 77881-538
3-Cyano-2-fluorobenzoic acid 97%
3-Cyano-2-fluorobenzoic acid

Precaución: For research use only.

Formula: C₈H₄FNO₂
MDL Number: MFCD09261249
CAS Number: 219519-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR