1-Bromo-2-chloro-4-(trifluoromethoxy)benzene 97%

Supplier: Ambeed
Warning

A105538-10G A105538-1G A105538-250MG A105538-25G A105538-5G
76970-398EA 25 USD
76970-398 76970-400 76970-402 76970-404 76970-406
1-Bromo-2-chloro-4-(trifluoromethoxy)benzene 97%
1-Bromo-2-chloro-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MDL Number: MFCD11111892
CAS Number: 892845-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR