1-Chloro-4-fluoro-2-methyl-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A192073-1G A192073-250MG A192073-25G A192073-5G
76974-874EA 11 USD
76974-874 76974-876 76974-878 76974-880
1-Chloro-4-fluoro-2-methyl-5-nitrobenzene 97%
1-Chloro-4-fluoro-2-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
MDL Number: MFCD11110548
CAS Number: 170098-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR