5'-Bromo-2'-methylacetanilide 98%

Supplier: Ambeed
Warning

A702348-1G A702348-5G
76981-104EA 21.99 USD
76981-104 76981-106
5'-Bromo-2'-methylacetanilide 98%
5'-Bromo-2'-methylacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MDL Number: MFCD11520466
CAS Number: 116436-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR