2-Fluoroterephthalic acid 95%

Supplier: Ambeed
Warning

A229434-1G A229434-5G A229434-250MG
76976-092EA 24 USD
76976-092 77550-488 77881-593
2-Fluoroterephthalic acid 95%
2-Fluoroterephthalic acid

Precaución: for Research Use Only

Formula: C₈H₅FO₄
MDL Number: MFCD03407274
CAS Number: 3906-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR