3-Aminopyrrolidine dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyrrolidin-3-amine dihydrochloride
A214163-100G
A214163-10G
A214163-1G
A214163-25G
A214163-5G
76975-606EA
199
USD
76975-606
76975-608
76975-610
76975-612
76975-614
3-Aminopyrrolidine dihydrochloride 97%
3-Aminopyrrolidine dihydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₀N₂ MW: 159.05 g/mol |
MDL Number:
MFCD00060176 CAS Number: 103831-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...