5-(1-(2,3-Dimethylphenyl)vinyl)-1H-imidazole 98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A386886-100MG A386886-250MG
76978-890EA 72.15 USD
76978-890 76978-892
5-(1-(2,3-Dimethylphenyl)vinyl)-1H-imidazole 98% stabilized
5-(1-(2,3-Dimethylphenyl)vinyl)-1H-imidazole

  • mix TBC as stabilizer


Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂
MW: 198.26 g/mol
MDL Number: MFCD13180466
CAS Number: 1021949-47-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR