Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 3a,4,7,7a-Tetrahydro-4,7-ethanoisobenzofuran-1,3-dione

A186698-1G A186698-25G A186698-5G
76974-672EA 18 USD
76974-672 76974-674 76974-676
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride 97%
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD06796361
CAS Number: 6708-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR