Methyl 3-amino-5-iodo-2-thiophenecarboxylate 98%

Supplier: Ambeed
Warning

A197886-10G A197886-1G A197886-250MG
76975-126EA 376.62 USD
76975-126 76975-128 76975-130
Methyl 3-amino-5-iodo-2-thiophenecarboxylate 98%
Methyl 3-amino-5-iodo-2-thiophenecarboxylate

Precaución: for Research Use Only

Formula: C₆H₆INO₂S
MW: 283.09 g/mol
MDL Number: MFCD18157627
CAS Number: 942589-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR