4-Bromo-5-ethoxyfuran-2(5H)-one 95%

Supplier: Ambeed
Warning

A263065-1G A263065-250MG A263065-5G A263065-25G
76977-012EA 22 USD
76977-012 76977-014 76977-016 77672-354
4-Bromo-5-ethoxyfuran-2(5H)-one 95%
4-Bromo-5-ethoxyfuran-2(5H)-one

Precaución: for Research Use Only

Formula: C₆H₇BrO₃
MDL Number: MFCD09953779
CAS Number: 32978-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR