4-Bromo-2-(trifluoromethoxy)benzonitrile 95%

Supplier: Ambeed
Warning

A203927-10G A203927-1G A203927-250MG A203927-25G A203927-5G
76975-302EA 97 USD
76975-302 76975-304 76975-306 76975-308 76975-310
4-Bromo-2-(trifluoromethoxy)benzonitrile 95%
4-Bromo-2-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrF₃NO
MDL Number: MFCD12761591
CAS Number: 1187983-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR