4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A492383-10G A492383-1G A492383-25G
76979-788EA 215.37 USD
76979-788 76979-790 76979-792
4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride 95%
4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O₂
MW: 202.6381 g/mol
MDL Number: MFCD18157694
CAS Number: 131020-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR