2'-Hydroxyacetophenone 98%

Supplier: Ambeed
Warning

A272252-1000G A272252-100G A272252-25G A272252-500G
76977-208EA 51 USD
76977-208 76977-210 76977-212 76977-214
2'-Hydroxyacetophenone 98%
2'-Hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₈O₂
MDL Number: MFCD00002219
CAS Number: 118-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR