Methyl 6-amino-2-chloronicotinate 96%

Supplier: Ambeed
Warning

A580278-250MG A580278-1G
76980-382EA 134.74 USD
76980-382 77594-762
Methyl 6-amino-2-chloronicotinate 96%
Methyl 6-amino-2-chloronicotinate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
MDL Number: MFCD14706237
CAS Number: 1004294-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR