2-(2-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)acetic acid 98%

Supplier: Ambeed
Warning

A157702-100MG A157702-250MG
76973-392EA 73.21 USD
76973-392 77611-508
2-(2-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)acetic acid 98%
2-(2-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD11100766
CAS Number: 7546-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR