(S)-2-(Pyrrolidin-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A174769-100MG A174769-1G A174769-250MG A174769-5G
76974-198EA 56.23 USD
76974-198 76974-200 76974-202 76974-204
(S)-2-(Pyrrolidin-2-yl)acetic acid 95%
(S)-2-(Pyrrolidin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD01863750
CAS Number: 56633-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR