4-Aminomethylpyridazine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A119674-100MG
A119674-1G
A119674-250MG
A119674-5G
76971-426EA
117
USD
76971-426
76971-428
76971-430
77880-968
4-Aminomethylpyridazine hydrochloride 97%
4-Aminomethylpyridazine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₈ClN₃ |
MDL Number:
MFCD16988465 CAS Number: 1351479-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...