2-Methylquinolin-4(1H)-one 97%

Supplier: Ambeed
Warning

A245841-1G A245841-250MG A245841-5G
76976-580EA 22 USD
76976-580 76976-582 77672-198
2-Methylquinolin-4(1H)-one 97%
2-Methylquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD00518775
CAS Number: 5660-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR