(R)-(+)-3-Benzyloxy-1,2-propanediol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-Glycerol alpha-Benzyl Ether, (R)-3-(Benzyloxy)propane-1,2-diol

A321328-100MG A321328-1G A321328-25G A321328-5G
76978-074EA 13.8 USD
76978-074 76978-076 76978-078 77672-648
(R)-(+)-3-Benzyloxy-1,2-propanediol 98%
(R)-(+)-3-Benzyloxy-1,2-propanediol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
MDL Number: MFCD00067260
CAS Number: 56552-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR