2-Chloro-4-(trifluoromethyl)benzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-(trifluoromethyl)benzylalcohol

A154606-10G A154606-1G A154606-250MG A154606-5G
76973-260EA 132.62 USD
76973-260 76973-262 76973-264 76973-266
2-Chloro-4-(trifluoromethyl)benzyl alcohol 97%
2-Chloro-4-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
MDL Number: MFCD04972757
CAS Number: 56456-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR