DL-Tryptophan methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride, H-DL-Trp-OMe·HCl

A140352-100G A140352-10G A140352-1G A140352-5G
76972-572EA 56.23 USD
76972-572 76972-574 76972-576 76972-578
DL-Tryptophan methyl ester hydrochloride 97%
DL-Tryptophan methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClN₂O₂
MW: 254.714 g/mol
MDL Number: MFCD00066133
CAS Number: 5619-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR