2-Bromo-4-methoxy-6-nitroaniline 98%

Supplier: Ambeed
Warning

A122596-10G A122596-1G A122596-5G
76971-634EA 181.42 USD
76971-634 76971-636 76971-638
2-Bromo-4-methoxy-6-nitroaniline 98%
2-Bromo-4-methoxy-6-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₃
MW: 247.046 g/mol
MDL Number: MFCD18088521
CAS Number: 10172-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR