Methyl-6-(hydroxymethyl)nicotinate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-(Hydroxymethyl)pyridine-3-carboxylate methyll ester, Methyl 6-(hydroxymethyl)nicotinate, 6-(Hydroxymethyl)nicotinic acid methyll ester, Methyl 6-(Hydroxymethyl)pyridine-3-carboxylate
A157303-1G
A157303-250MG
A157303-5G
A157303-10G
A157303-25G
76973-364EA
24.41
USD
76973-364
76973-366
76973-368
77671-232
77671-234
Methyl-6-(hydroxymethyl)nicotinate 97%
Methyl-6-(hydroxymethyl)nicotinate
Precaución: for Research Use Only
Formula:
C₈H₉NO₃ MW: 167.1600 g/mol |
MDL Number:
MFCD00643821 CAS Number: 56026-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...