2-Amino-4-chloro-6-methylpyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-6-methylpyrimidin-2-amine

A105136-100G A105136-10G A105136-25G A105136-5G
76970-386EA 108.22 USD
76970-386 76970-388 76970-390 77570-308
2-Amino-4-chloro-6-methylpyrimidine 98%
2-Amino-4-chloro-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD00006091
CAS Number: 5600-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR