3,4-Dichloropyridine 97%

Supplier: Ambeed
Warning

A120247-25G A120247-100G A120247-5G
76971-464EA 45 USD
76971-464 77670-708 77880-971
3,4-Dichloropyridine 97%
3,4-Dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N
MDL Number: MFCD01861989
CAS Number: 55934-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR