Ethyl N-(4-methylbenzenesulfonyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl tosylcarbamate

A210482-100MG A210482-10G A210482-250MG A210482-5G A210482-1G
76975-522EA 5.31 USD
76975-522 76975-524 76975-526 76975-528 77612-068
Ethyl N-(4-methylbenzenesulfonyl)carbamate 95%
Ethyl N-(4-methylbenzenesulfonyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄S
MW: 243.27 g/mol
MDL Number: MFCD00797861
CAS Number: 5577-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR