2-(Piperazin-1-yl)-1H-benzo[d]imidazole hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A766862-100MG
A766862-1G
A766862-250MG
A766862-50MG
A766862-5G
76981-460EA
83
USD
76981-460
76981-462
76981-464
76981-466
76981-468
2-(Piperazin-1-yl)-1H-benzo[d]imidazole hydrochloride 95%
2-(Piperazin-1-yl)-1H-benzo[d]imidazole hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₅ClN₄ MW: 238.71 g/mol |
MDL Number:
MFCD09877692 CAS Number: 1185310-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...