2-Chloro-5-(ethoxymethyl)pyridine 95%

Supplier: Ambeed
Warning

A351870-100MG A351870-1G A351870-250MG
76978-516EA 24.41 USD
76978-516 76978-518 76978-520
2-Chloro-5-(ethoxymethyl)pyridine 95%
2-Chloro-5-(ethoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD09808844
CAS Number: 871829-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR