Trimesic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid
A284402-100G
A284402-25G
A284402-500G
A284402-1000G
76977-482EA
9.55
USD
76977-482
76977-484
77550-746
77571-768
Trimesic acid 98%
Trimesic acid
Precaución: for Research Use Only
Formula:
C₉H₆O₆ MW: 210.14 g/mol |
MDL Number:
MFCD00002517 CAS Number: 554-95-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...