2,4-Dichloro-5-(ethoxymethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A169271-1G A169271-5G
76973-908EA 57.29 USD
76973-908 76973-910
2,4-Dichloro-5-(ethoxymethyl)pyrimidine 97%
2,4-Dichloro-5-(ethoxymethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂O
MW: 207.05 g/mol
MDL Number: MFCD18837148
CAS Number: 7627-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR