2,6-Bis((R)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A387545-1G A387545-250MG A387545-100MG
76978-898EA 454 USD
76978-898 76978-900 77882-035
2,6-Bis((R)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((R)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₄₁H₃₉N₃O₂
MDL Number: MFCD18379148
CAS Number: 828918-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR