3-(Aminomethyl)cyclohexanol 95%

Supplier: Ambeed
Warning

A393326-1G A393326-5G
76978-982EA 63 USD
76978-982 76978-984
3-(Aminomethyl)cyclohexanol 95%
3-(Aminomethyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MDL Number: MFCD07786652
CAS Number: 116650-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR