6-Amino-3-bromo-2-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A120820-100MG A120820-10MG A120820-1G A120820-250MG A120820-5G
76971-504EA 48.81 USD
76971-504 76971-506 76971-508 76971-510 76971-512
6-Amino-3-bromo-2-methylbenzoic acid 95%
6-Amino-3-bromo-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.05 g/mol
MDL Number: MFCD13659393
CAS Number: 124341-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR